Learn More
Electron Microscopy Sciences CONICALCNTRFGE TBE 50ML
SDP
Catalog No. 502413126
Encompass_Preferred
EzFlip Conical Centrifuge tubes offers a safer alternative to screw cap tubes. A patented closure system improves ergonomics and versatility. An improved tube and cap design increases mechanical strength to withstand aggressive centrifugal forces. The 15 ml tube's maximum relative centrifugal force (RCF) is 8,500 x g, and the 50 ml tube's maximum RCF is 9,500 x g. Tubes are graduated and feature a writing area.
Catalog No. 50-241-3126 Supplier Electron Microscopy Sciences Supplier No. 6303125
EzFlip Conical Centrifuge tubes offers a safer alternative to screw cap tubes. A patented closure system improves ergonomics and versatility. An improved tube and cap design increases mechanical strength to withstand aggressive centrifugal forces. The 15 ml tube's maximum relative centrifugal force (RCF) is 8,500 x g, and the 50 ml tube's maximum RCF is 9,500 x g. Tubes are graduated and feature a writing area.