Learn More
Medchemexpress LLC Coniferaldehyde | 458-36-6 | 100 MG
SDP
Catalog No. 5000418238
Encompass_Preferred
Coniferaldehyde (4-Hydroxy-3-methoxycinnamaldehyde) is an effective inducer of heme oxygenase-1 (HO-1). It inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway in RAW264.7 macrophage cells, and possesses antioxidant and anti-inflammatory activities.
  • Effective inducer of heme oxygenase-1 (HO-1).
  • Inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway in RAW264.7 macrophage cells.
  • Possesses antioxidant activities.
  • Possesses anti-inflammatory activities.
  • Activates the Nrf2 signaling pathway in primary chondrocytes and articular cartilage from the knee joints.
  • Alleviates cartilage destruction in OA mice.
Catalog No. 50-004-18238 Supplier Medchemexpress LLC Supplier No. HYN2535100MG
Coniferaldehyde (4-Hydroxy-3-methoxycinnamaldehyde) is an effective inducer of heme oxygenase-1 (HO-1). It inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway in RAW264.7 macrophage cells, and possesses antioxidant and anti-inflammatory activities.
  • Effective inducer of heme oxygenase-1 (HO-1).
  • Inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway in RAW264.7 macrophage cells.
  • Possesses antioxidant activities.
  • Possesses anti-inflammatory activities.
  • Activates the Nrf2 signaling pathway in primary chondrocytes and articular cartilage from the knee joints.
  • Alleviates cartilage destruction in OA mice.