Learn More
Medchemexpress LLC Coniferaldehyde | 458-36-6 | 50 MG
SDP
Catalog No. 5000420920
Encompass_Preferred
Coniferaldehyde (4-Hydroxy-3-methoxycinnamaldehyde) is a chemical compound known for its ability to induce heme oxygenase-1 (HO-1). It demonstrates antioxidant and anti-inflammatory activities and inhibits LPS-induced apoptosis in RAW264.7 macrophage cells.
  • Effective inducer of heme oxygenase-1 (HO-1).
  • Inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway.
  • Possesses antioxidant activities.
  • Exhibits anti-inflammatory activities.
Catalog No. 50-004-20920 Supplier Medchemexpress LLC Supplier No. HYN253550MG
Coniferaldehyde (4-Hydroxy-3-methoxycinnamaldehyde) is a chemical compound known for its ability to induce heme oxygenase-1 (HO-1). It demonstrates antioxidant and anti-inflammatory activities and inhibits LPS-induced apoptosis in RAW264.7 macrophage cells.
  • Effective inducer of heme oxygenase-1 (HO-1).
  • Inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway.
  • Possesses antioxidant activities.
  • Exhibits anti-inflammatory activities.