Learn More
Medchemexpress LLC HY-N2535 5mg Medchemexpress, Coniferaldehyde CAS:458-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25355MG
Encompass_Preferred
Medchemexpress, HY-N2535 5mg Coniferaldehyde CAS:458-36-6 Coniferaldehyde (Ferulaldehyde), isolated from the ethanol extract of Vitex rotundifolia fruits, is an effective inducer of heme oxygenase-1 (HO-1). Coniferaldehyde exerts anti-inflammatory properties in response to LPS. Coniferaldehyde inhibits LPS-induced apoptosis through the PKCα/β II/Nrf-2/HO-1 dependent pathway in RAW264.7 macrophage cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0402