Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Coniferyl alcohol

Catalog No. 223735100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-373-5100MG 100 mg
22-373-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-373-5100MG Supplier Sigma Aldrich Supplier No. 223735100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Coniferyl alcohol

Specifications

CAS 458-35-5
Melting Point 75°C to 80°C (lit.)
Boiling Point 163°C to 165°C (3 mmHg)
Molecular Formula C10H12O3
Linear Formula HOC6H3(OCH3)CH=CHCH2OH
MDL Number MFCD00002922
Quantity 100 mg
Synonym 3-(4-Hydroxy-3-methoxyphenyl)-2-propen-1-ol; 4-Hydroxy-3-methoxycinnamyl alcohol
Molecular Weight (g/mol) 180.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.