Learn More
Medchemexpress LLC Coniferyl alcohol | 458-35-5 | 50 MG
SDP
Catalog No. 5000418096
Encompass_Preferred
Coniferyl alcohol is an orally active intermediate in lignin biosynthesis and functions as an antifungal agent. It has been shown to improve cardiac dysfunction in renovascular hypertension and cardiac inflammation.
  • Inhibits the growth of fungi such as Verticillium longisporum
  • Inhibits the growth of Nicotiana benthamiana seedlings
  • Alleviates cardiac inflammation and hypertrophy in renovascular hypertension models
  • Reduces systolic and diastolic blood pressures in renovascular hypertension models
Catalog No. 50-004-18096 Supplier Medchemexpress LLC Supplier No. HYN428350MG
Coniferyl alcohol is an orally active intermediate in lignin biosynthesis and functions as an antifungal agent. It has been shown to improve cardiac dysfunction in renovascular hypertension and cardiac inflammation.
  • Inhibits the growth of fungi such as Verticillium longisporum
  • Inhibits the growth of Nicotiana benthamiana seedlings
  • Alleviates cardiac inflammation and hypertrophy in renovascular hypertension models
  • Reduces systolic and diastolic blood pressures in renovascular hypertension models