Learn More
Selleck Chemical LLC Coniferyl alcohol
SDP
Catalog No. 5000004730
Encompass_Preferred
Coniferyl alcohol is an intermediate in biosynthesis of eugenol and of stilbene and coumarin
Catalog No. 50-000-04730 Supplier Selleck Chemical LLC Supplier No. S64295MG
Coniferyl alcohol is an intermediate in biosynthesis of eugenol and of stilbene and coumarin