Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coniferyl alcohol, 98%

Catalog No. p-7027891
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2494901 0.25 g
AAB2494903 1 g
AAB2494906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2494901 Supplier Thermo Scientific Chemicals Supplier No. B2494901
Only null left
Add to Cart

CAS: 458-35-5 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-35-5
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00002922
InChI Key JMFRWRFFLBVWSI-NSCUHMNNSA-N
Synonym coniferyl alcohol, coniferol, coniferylalcohol, 4-3-hydroxyprop-1-en-1-yl-2-methoxyphenol, gamma-hydroxyisoeugenol, trans-coniferyl alcohol, e-coniferyl alcohol, 4-3-hydroxy-1-propenyl-2-methoxyphenol, 3-4-hydroxy-3-methoxyphenyl-2-propen-1-ol, 4-1e-3-hydroxyprop-1-en-1-yl-2-methoxyphenol
PubChem CID 1549095
ChEBI CHEBI:17745
SMILES COC1=CC(\C=C\CO)=CC=C1O

Specifications

Melting Point 75°C to 76°C
Boiling Point 163°C to 165°C (3 mmHg)
Quantity 0.25 g
Beilstein 2048961
Sensitivity Air and light sensitive
Merck Index 14,2504
IUPAC Name 4-[(1E)-3-hydroxyprop-1-en-1-yl]-2-methoxyphenol
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Coniferyl alcohol
EINECSNumber 207-277-9
RTECSNumber SL5367000
TSCA Yes
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.