Learn More
Cayman Chemical Coniferyl Ferulate, Purity: ≥95%, CAS: 63644-62-2, 10 mg
SDP
Catalog No. 502991993
Encompass_Preferred
A monolignol conjugate; active against B. subtilis and S. aureus; scavenges DPPH radicals (EC50 = 3.6 µg/ml) and inhibits GST (IC50 = 0.3 µM for the human placental enzyme) in cell-free assays; reduces cytotoxicity induced by Aβ40 in PC12 cells at 10 and 50 µg/ml
Catalog No. 50-299-1993 Supplier Cayman Chemical Supplier No. 2962010 MG
A monolignol conjugate; active against B. subtilis and S. aureus; scavenges DPPH radicals (EC50 = 3.6 µg/ml) and inhibits GST (IC50 = 0.3 µM for the human placental enzyme) in cell-free assays; reduces cytotoxicity induced by Aβ40 in PC12 cells at 10 and 50 µg/ml