Learn More
Medchemexpress LLC HY-N1916 5mg Medchemexpress, Coniferyl ferulate CAS:63644-62-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19165MG
Encompass_Preferred
Medchemexpress, HY-N1916 5mg Coniferyl ferulate CAS:63644-62-2 Coniferyl ferulate, a strong inhibitor of glutathione S-transferase (GST) isolated from Radix Angelicae sinensis, reverses multidrug resistance and downregulates P-glycoprotein. Coniferyl ferulate shows strong inhibition of human placental GST with an IC50 of 0.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2852