Learn More
Medchemexpress LLC -Coniine hydroch 50mg
SDP
Catalog No. 5000205479
Encompass_Preferred
( )-Coniine hydrochloride (2-Propylpiperidine hydrochloride) is a potent nAChR agonist with an EC50 value of 0 3 mM ( )-Coniine hydrochloride shows acute toxicity with an LD50 value of 7 7 mg/kg[1]
Catalog No. 50-002-05479 Supplier Medchemexpress LLC Supplier No. HYW09975750MG
( )-Coniine hydrochloride (2-Propylpiperidine hydrochloride) is a potent nAChR agonist with an EC50 value of 0 3 mM ( )-Coniine hydrochloride shows acute toxicity with an LD50 value of 7 7 mg/kg[1]