Learn More
Glass Expansion Conikal DC Nebulizer 2mL/min suitable for Thermo Pro, 6000 series and 7000 series ICP-OES. Includes NFT-075 UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 piece) and Thermo DC gas line 70-803-1292.
SDP
Catalog No. 502703013
Encompass_Preferred
Conikal DC Nebulizer 2.0mL/min, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 31 with ratchet connector (1 pc). Compatible with Thermo Pro, 6000 Series and 7000 Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 2.0mL/min. Recommended not to exceed optimum 2.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates.Replacement parts include P/Ns NC0277960 and 501907300.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3013 Supplier Glass Expansion Supplier No. A3107UC2
Conikal DC Nebulizer 2.0mL/min, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 31 with ratchet connector (1 pc). Compatible with Thermo Pro, 6000 Series and 7000 Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 2.0mL/min. Recommended not to exceed optimum 2.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates.Replacement parts include P/Ns NC0277960 and 501907300.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001