Learn More
Glass Expansion Conikal DC Nebulizer 1mL/min
SDP
Catalog No. 5000556396
Encompass_Preferred
Conikal DC Nebulizer 1mL/min Sourced from Glass Expansion
Catalog No. 50-005-56396 Supplier Glass Expansion Supplier No. A1305UC1
Conikal DC Nebulizer 1mL/min Sourced from Glass Expansion