Learn More
Glass Expansion Conikal DC Nebulizer 1mL/min, Includes NFT-16-50 UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece) and 70-803-0969 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907361
Encompass_Preferred
Conikal DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Vista/700-ES Series ICP-OES, 4100/4200 MP-AES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates.Replacement parts include P/Ns 502702959 and 502703076.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7361 Supplier Glass Expansion Supplier No. A1107UC1
Conikal DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Vista/700-ES Series ICP-OES, 4100/4200 MP-AES.Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates.Replacement parts include P/Ns 502702959 and 502703076.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001