Learn More
Glass Expansion Conikal Nebulizer 1mL/min with EzyFit & EzyLok, Includes EZTS-16-75 EzyFit Connector with 0.75mm ID x 1.6mm OD x 1500mm L tubing (1 unit)
SDP
Catalog No. 501907495
Encompass_Preferred
Conikal Nebulizer 1.0 mL/min, includes EzyFit Connector with 0.75 mm ID x 1.6 mm OD x 1500 mm long tubing (1 piece) and EzyLok Connector (1 piece).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 1.0 mL/min. Recommended not to exceed optimum 1.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. Backpressure should read approximately 30psi at 1.0 L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates. Replacement parts include P/Ns EZTS-16-75 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) Conikal to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7495 Supplier Glass Expansion Supplier No. AR301FC1ES
Conikal Nebulizer 1.0 mL/min, includes EzyFit Connector with 0.75 mm ID x 1.6 mm OD x 1500 mm long tubing (1 piece) and EzyLok Connector (1 piece).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 1.0 mL/min. Recommended not to exceed optimum 1.0 mL/min sample flow rate; however, lower gas and sample flow rates are OK. Backpressure should read approximately 30psi at 1.0 L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates. Replacement parts include P/Ns EZTS-16-75 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) Conikal to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001