Learn More
Glass Expansion Conikal U-Series Nebulizer 1mL/min, Includes NFT-16-50 UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece), and EL-1 EzyLok Connector Kit.
SDP
Catalog No. 501907521
Encompass_Preferred
Conikal U-Series Nebulizer 1.0 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and EzyLok Connector (1 pc.).Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates. Replacement parts include P/Ns 502702959 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) Conikal to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7521 Supplier Glass Expansion Supplier No. ARG07UC1
Conikal U-Series Nebulizer 1.0 mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc.) and EzyLok Connector (1 pc.).Optimum nebulizer gas flow rate is 0.7L/min and sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40psi at 0.7L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates. Replacement parts include P/Ns 502702959 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) Conikal to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001