Learn More
Perkin Elmer US LLC Conikal U-Series Nebulizer 2 mL/min with EzyFit
Catalog No. 502104200
Encompass_Preferred
Includes UniFit Connector with 0.75 mm I.D. x 1.3 mm O.D. x 700 mm long tubing (N0777512) and EzyLok Connector Kit (N0777413).
Catalog No. 50-210-4200 Supplier Perkin Elmer US LLC Supplier No. N0777487
Includes UniFit Connector with 0.75 mm I.D. x 1.3 mm O.D. x 700 mm long tubing (N0777512) and EzyLok Connector Kit (N0777413).