Learn More
Selleck Chemical LLC Conivaptan HCl 10mg 168626-94-6
SDP
Supplier:  Selleck Chemical LLC S211610MG
Encompass_Preferred
Conivaptan HCl is an orally active, non-peptide, vasopressin V1A and V2 receptor antagonist, used in the treatment of euvolemic and hypervolemic hyponatremia. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2073