Learn More
Apexbio Technology LLC Conivaptan HCl 168626-94-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093932
Encompass_Preferred
Conivaptan Hydrochloride (CAS 168626-94-6) is a non-peptide antagonist of the antidiuretic hormone vasopressin It selectively inhibits vasopressin V1A and V2 receptors thereby blocking vasopressin-mediated water reabsorption in renal collecting ducts This action promotes aquaresis without affecting sodium or potassium excretion Conivaptan Hydrochloride is utilized in biomedical research to investigate pathways involved in fluid balance sodium homeostasis and cardiovascular dysfunction including models of congestive heart failure and disorders of hyponatremia
Catalog No. 50-000-93932 Supplier Apexbio Technology LLC Supplier No. A84025.1
Conivaptan Hydrochloride (CAS 168626-94-6) is a non-peptide antagonist of the antidiuretic hormone vasopressin It selectively inhibits vasopressin V1A and V2 receptors thereby blocking vasopressin-mediated water reabsorption in renal collecting ducts This action promotes aquaresis without affecting sodium or potassium excretion Conivaptan Hydrochloride is utilized in biomedical research to investigate pathways involved in fluid balance sodium homeostasis and cardiovascular dysfunction including models of congestive heart failure and disorders of hyponatremia