Learn More
Medchemexpress LLC Conivaptan hydrochloride | 168626-94-6 | 99.9% | 535.04 | 1 ML
SDP
Catalog No. 5000387699
Encompass_Preferred
Conivaptan hydrochloride is a non-peptide antagonist of the vasopressin receptor, demonstrating Ki values of 0.48 nM for the rat liver V1A receptor and 3.04 nM for the rat kidney V2 receptor.
  • Increases urine volume and reduces urine osmolality.
  • Reduces right ventricular systolic pressure and left ventricular end-diastolic pressure.
  • Normalizes urinary sodium excretion without affecting creatinine clearance or arterial pressure.
  • Exerts dose-dependent diuretic effects without increasing electrolyte excretion.
  • Improves cardiac function in congestive heart failure models.
  • For research use only.
Catalog No. 50-003-87699 Supplier Medchemexpress LLC Supplier No. HY18347A10MM/1ML
Add to Cart
Add to Cart
Conivaptan hydrochloride is a non-peptide antagonist of the vasopressin receptor, demonstrating Ki values of 0.48 nM for the rat liver V1A receptor and 3.04 nM for the rat kidney V2 receptor.
  • Increases urine volume and reduces urine osmolality.
  • Reduces right ventricular systolic pressure and left ventricular end-diastolic pressure.
  • Normalizes urinary sodium excretion without affecting creatinine clearance or arterial pressure.
  • Exerts dose-dependent diuretic effects without increasing electrolyte excretion.
  • Improves cardiac function in congestive heart failure models.
  • For research use only.