Learn More
Medchemexpress LLC Conivaptan hydrochloride | 168626-94-6 | 99.95% | 535.04 | 100 MG
SDP
Catalog No. 5000387545
Encompass_Preferred
Conivaptan hydrochloride is a non-peptide antagonist of the vasopressin receptor, demonstrating high affinity for both V1A and V2 receptors. It is intended for research use only and not for medical applications. This compound exhibits significant biological activity, including increasing urine volume and reducing urine osmolality in myocardial infarction rats, and improving cardiac function in dogs with congestive heart failure.
  • Non-peptide vasopressin receptor antagonist
  • High affinity for V1A and V2 receptors
  • Increases urine volume and reduces urine osmolality
  • Improves cardiac function in relevant models
  • Suitable for in vitro and in vivo research studies
Catalog No. 50-003-87545 Supplier Medchemexpress LLC Supplier No. HY18347A100MG
Conivaptan hydrochloride is a non-peptide antagonist of the vasopressin receptor, demonstrating high affinity for both V1A and V2 receptors. It is intended for research use only and not for medical applications. This compound exhibits significant biological activity, including increasing urine volume and reducing urine osmolality in myocardial infarction rats, and improving cardiac function in dogs with congestive heart failure.
  • Non-peptide vasopressin receptor antagonist
  • High affinity for V1A and V2 receptors
  • Increases urine volume and reduces urine osmolality
  • Improves cardiac function in relevant models
  • Suitable for in vitro and in vivo research studies