Learn More
TARGETMOL CHEMICALS INC CONIVAPTAN HYDROCHLORIDE 25MG
SDP
Catalog No. 502576515
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Conivaptan hydrochloride (Vaprisol) is an orally active vasopressin V2 and V1A receptor antagonist used in the therapy of hypervolemic hyponatremia and euvolemic. purity: 99%
Catalog No. 50-257-6515 Supplier TARGETMOL CHEMICALS INC Supplier No. T645325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Conivaptan hydrochloride (Vaprisol) is an orally active vasopressin V2 and V1A receptor antagonist used in the therapy of hypervolemic hyponatremia and euvolemic. purity: 99%