Learn More
Cayman Chemical Conivaptan-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1129433-63-1, 1 mg
SDP
Catalog No. 502935443
Encompass_Preferred
An internal standard for the quantification of conivaptan by GC- or LC-MS
Catalog No. 50-293-5443 Supplier Cayman Chemical Supplier No. 264551 MG
An internal standard for the quantification of conivaptan by GC- or LC-MS