Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conjugated (10E,12Z)-Linoleic acid Solution, 100 mg/mL in ethanol, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018659 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8659 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8659 Supplier MilliporeSigma Supelco Supplier No. 0439750MG
Add to Cart
Add to Cart

Specifications

CAS 2420-56-6
Physical Form Liquid
Linear Formula C18H32O2
MDL Number MFCD02259040
Synonym (10 E,12 Z)-10,12-Octadecadienoic acid; 10 E,Z12-CLA; Linoleic acid (10-trans, 12-cis)
Type Analytical Standard
Quantity 50 mg
Formula Weight 280.45
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 mg/mL in ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.