Learn More
MOLECULAR DEPOT LLC Conjugated Bilirubin - 1 mL
Catalog No. 5000013454
Encompass_Preferred
Conjugated Bilirubin for Interference Testing a highly concentrated solution of conjugated bilirubin (20 mg/mL) provided with the corresponding diluent This solution can be spiked into specimens to test conjugated bilirubin interference Instructions with sample calculations are also provided For in vitro research use only
Catalog No. 50-000-13454 Supplier MOLECULAR DEPOT LLC Supplier No. K2010001C
Conjugated Bilirubin for Interference Testing a highly concentrated solution of conjugated bilirubin (20 mg/mL) provided with the corresponding diluent This solution can be spiked into specimens to test conjugated bilirubin interference Instructions with sample calculations are also provided For in vitro research use only