Learn More
Cayman Chemical 10(E),12(Z)-Conjugated Linoleic Acid, 2420-56-6, 10 mg
SDP
Supplier:  Cayman Chemical 9014510
Encompass_Preferred
CLA refers to a family of 8 geometric isomers of linoleic acid in which the two double bonds are contiguous. (The predominant form of linoleic acid in nature, 18:ω6, has double bonds at 9 and 12, interrupted by a methylene carbon.) CLA is found in both meat and dairy products, but it is not found to any significant degree in plants. The predominant isomer of CLA in animal tissue is 9Z,11E; smaller amounts of 10E,12Z also occur. Various antioxidant and antitumor activities have been attributed to CLA or its downstream metabolites.{6849} Reported health benefits of dietary supplementation with CLA have been attributed variously to competitive inhibition of .DELTA.6-desaturase and/or PPARγ activation.{6980}
Catalog No. NC0870014