Learn More
Vector Laboratories Conjugation Buffer (10X), 5 x 1.5 mL
SDP
Catalog No. 502045756
Encompass
Bioconjugation. Conjugation Buffer is a high buffering capacity solution which is optimized for the conjugation reaction between HyNic and 4FB groups. Proteins, oligonucleotides, and other biomolecules are buffer exchanged into Conjugation Buffer after modification with S-HyNic and S-4FB, and prior to conjugation, to ensure efficient conjugation of these linkers.
Catalog No. 50-204-5756 Supplier Vector Laboratories Supplier No. S4002005
Bioconjugation. Conjugation Buffer is a high buffering capacity solution which is optimized for the conjugation reaction between HyNic and 4FB groups. Proteins, oligonucleotides, and other biomolecules are buffer exchanged into Conjugation Buffer after modification with S-HyNic and S-4FB, and prior to conjugation, to ensure efficient conjugation of these linkers.