Learn More
Ace Glass, Inc. Cord, Instatherm, banana jack type, 6ft, fits pre-2009 baths
SDP
Catalog No. 509558886
Encompass
6 Instatherm connecting cord features in-line fuse protection against surges and high voltage. Cord has banana jack type plug for use with pre-2009 Instatherm baths.
Catalog No. 50-955-8886 Supplier Ace Glass, Inc. Supplier No. 969816
6 Instatherm connecting cord features in-line fuse protection against surges and high voltage. Cord has banana jack type plug for use with pre-2009 Instatherm baths.