Learn More
Ace Glass, Inc. CONN FOR -26 CONN FOR -26
SDP
Supplier:  Ace Glass, Inc. 781828
Encompass_Preferred
Surge Flask Connector with 4mm stopcock for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-464