Learn More
LumiraDx Connect Hub
Catalog No. 22181003 Shop All Roche Diagnostics Products
Change view
Click to view available options
Type:
Wired or WiFi Network Access Point
Catalog No. Type
22-181-003 Wired or WiFi Network Access Point
1 options
Catalog No. 22-181-003 Supplier LumiraDx Supplier No. 10417889001
Add to Cart
Add to Cart
Wired or WiFi network access point to enable secure bidirectional data exchange to and from the LumiraDx instrument
  • Features:
    • Wired or WiFi network access point to enable secure bidirectional data exchange to and from the LumiraDx Instrument as part of the LumiraDx Platform
    • One connect hub can wirelessly communicate with up to seven LumiraDx instruments at one time
TRUSTED_SUSTAINABILITY
Specifications
Type Wired or WiFi Network Access Point