Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connecticut Clean Room™ Mats

Catalog No. 19041570
Change view
Click to view available options
Dimensions (L x W):
24 x 45 in.
26 x 45 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-041-570 24 x 45 in.
19-041-569 26 x 45 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-041-570 Supplier Connecticut Clean Room™ Supplier No. P112B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Removes footborne particulates before entering cleanroom

For use before entering cleanrooms or controlled areas to remove foot borne particulate, including dirt, dust and other foreign debris.

  • Seamless one-piece construction
  • Made from incinerable polyethylene film
  • Contains special anti-microbial agent providing long-lasting protection against bacterial growth
  • Numbered tabs provide easy identification and sheet separation
  • Easy mobility for carts and wheeled machinery

Compliance

Adhesive level minimum of 280.95mL (9.5 oz.) per square inch with stainless steel method per PSTC-1 (Pressure Sensitive Tape Council).

Specifications

Quantity 30 Sheets/Mat
For Use With (Application) Removing dirt and dust from foot traffic and equipment wheels
Dimensions (L x W) 24 x 45 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.