Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connecticut Clean Room™ Tyvek™ Coveralls

Catalog No. 19808712
Change view
Click to view available options
:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-808-712 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-808-712 Supplier Connecticut Clean Room™ Supplier No. DCWHLG
This item is not returnable. View return policy

Specifications

Product Type Cleanroom Coveralls
Size Large
Material Tyvek
Color White
Disposable Single-use
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.