Learn More
MSC Value Collection Roller Chain Link, ANSI 80, Carbon Steel, 10/Pk.
Catalog No. 5000514537
Encompass
This roller chain link is designed for use with single strand chains, providing a secure connection for mechanical power transmission systems. Constructed from carbon steel, this component offers the durability required for industrial chain applications.
  • Compatible with ANSI 80 single strand roller chain
  • Constructed from carbon steel for increased durability
  • Designed for reliable chain connection and performance
Catalog No. 50-005-14537 Supplier MSC Supplier No. 53613964
This roller chain link is designed for use with single strand chains, providing a secure connection for mechanical power transmission systems. Constructed from carbon steel, this component offers the durability required for industrial chain applications.
  • Compatible with ANSI 80 single strand roller chain
  • Constructed from carbon steel for increased durability
  • Designed for reliable chain connection and performance