Learn More
Chemglass Life Sciences Tube, Connecting, "T" Shaped, CG-300-01 (9mm Small Serration, 10mm Large Serration) Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG402201
Encompass_Preferred
Tube, Connecting, "T" Shaped, CG-300-01 (9mm Small Serration, 10mm Large Serration) Hose Connection. T shaped tube having serrated ends for connection of rubber, latex or Tygon tubing.
Catalog No. CG402201