Learn More
Chemglass Life Sciences Thread size: GL-14, Style: Straight, Hose OD (mm): 8.7, Hose ID (mm): 5, Length (mm): 25
SDP
Catalog No. 50872170
Encompass_Preferred
Removable hose connection, Thread size: GL-14, Style: Straight, Hose OD (mm): 8.7, Hose ID (mm): 5, Length (mm): 25. Polypropylene hose connections with a Viton rubber seal for use with GL-14 glass screw threads for working temperatures up to max +110¿C. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing Tygon tubing over a serrated glass connector.
Catalog No. 50-872-170 Supplier Chemglass Life Sciences Supplier No. CG19802
Removable hose connection, Thread size: GL-14, Style: Straight, Hose OD (mm): 8.7, Hose ID (mm): 5, Length (mm): 25. Polypropylene hose connections with a Viton rubber seal for use with GL-14 glass screw threads for working temperatures up to max +110¿C. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing Tygon tubing over a serrated glass connector.