Learn More
Chemglass Life Sciences Thread size: GL-18, Style: Straight, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 51
SDP
Catalog No. 50872172
Encompass_Preferred
Removable hose connection, Thread size: GL-18, Style: Straight, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 51. Polypropylene hose connections with a Viton rubber seal for use with GL-18 glass screw threads for working temperatures up to max +110¿C. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing Tygon tubing over a serrated glass connector.
Catalog No. 50-872-172 Supplier Chemglass Life Sciences Supplier No. CG19805
Removable hose connection, Thread size: GL-18, Style: Straight, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 51. Polypropylene hose connections with a Viton rubber seal for use with GL-18 glass screw threads for working temperatures up to max +110¿C. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing Tygon tubing over a serrated glass connector.