Learn More
Binder Connection Kit VDL VP5 PTFE
SDP
Catalog No. 11103190 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With:
Vacuum Pump VP5 EX
Catalog No. For Use With
11-103-190 Vacuum Pump VP5 EX
1 options
Catalog No. 11-103-190 Supplier Binder Supplier No. 80122614
Add to Cart
Add to Cart
For vacuum pump VP5 EX.
  • Consisting of antistatic PTFE hose 1.9 m long, with small flange 1/2 in. in diameter (DN 16) on both sides, two clamping rings and two universal centering rings.
Specifications
Autoclavable Not Autoclavable
For Use With Vacuum Pump VP5 EX
IP Rating IP52
Type Pump Connection Kit