Learn More
Binder Connection Kit VDL VP5 Stainless Steel
SDP
Catalog No. 11103197 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With:
VP5 Chemical Membrane Pump
Catalog No. For Use With
11-103-197 VP5 Chemical Membrane Pump
1 options
Catalog No. 11-103-197 Supplier Binder Supplier No. 80122724
Add to Cart
Add to Cart
For vacuum pump VP5 EX.
  • Consisting of stainless steel hose 1.9 m long, with small flange 1/2 in. in diameter (DN 16) on both sides, two clamping rings and two universal centering rings.
Specifications
For Use With VP5 Chemical Membrane Pump
IP Rating IP52
Tubing Size DN 16
Type Pump Connection Kit