Learn More
Harvard Apparatus™ Connection Kit for VentElite for Pressurized Gas
SDP
Catalog No. 14516261
Change view
Click to view available options
Animal:
Mouse, Rat, Guinea Pig
Catalog No. Animal
14-516-261 Mouse, Rat, Guinea Pig
1 options
Catalog No. 14-516-261 Supplier Harvard Apparatus™ Supplier No. 734872
Add to Cart
Add to Cart
Necessary for connection of your small animal ventilator to anesthesia system.
  • Directly connect anesthesia to ventilator for appropriate delivery to intubated or tracheotomized animal.
  • Removes pressurized gas to waste port preventing lung and ventilator damage.
  • Can also be used to safely connect any pressurized gas source to the inlet of a ventilator.
  • Several sizes available to work with all Harvard Apparatus Ventilators.
Specifications
Animal Mouse, Rat, Guinea Pig
Type Connection Kits
Disposable Single-use
For Use With (Equipment) VentElite, Model 683, MicroVent, MiniVent, MidiVent
Includes Connection Kit for Pressurized Gas