Learn More
Perkin Elmer US LLC Flow Injection Connector with 1⁄4 in. (6.4 mm) Internal Screw Thread, Type 2
Catalog No. 509042331
Encompass_Preferred
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. Type 2 in instrument manual diagrams.
Catalog No. 50-904-2331 Supplier Perkin Elmer US LLC Supplier No. B0196704
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. Type 2 in instrument manual diagrams.