Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316 Stainless Steel, Straight, Hose Barb Reducer, 1/8"" ID x 1/16"" ID; Each
Catalog No. 50629430
Encompass
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilize via ethylene oxide, autoclave, or gamma irradiation.
Catalog No. 50-629-430 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062009
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilize via ethylene oxide, autoclave, or gamma irradiation.