Learn More
Grainger Connector 1 CTS x 3/4 NPT White
Catalog No. 502767349
Encompass_Preferred
The John Guest Male Connector is a plastic push-to-connect style fitting for use with polyethylene tubing. It has a 1/4" NPT male thread and is suitable for pressures up to 232 PSI. The body is made from acetal copolymer plastic. It connects to tubing sizes from 1/4" to 5/16" outer diameter. This fitting provides a quick and easy way to connect polyethylene tubing to threaded NPT ports. It can handle a variety of fluids including air, water, and chemicals. The push-to-connect design allows tubing to be securely connected without requiring clamps or threads.
Catalog No. 50-276-7349 Supplier Grainger Supplier No. 16T761
The John Guest Male Connector is a plastic push-to-connect style fitting for use with polyethylene tubing. It has a 1/4" NPT male thread and is suitable for pressures up to 232 PSI. The body is made from acetal copolymer plastic. It connects to tubing sizes from 1/4" to 5/16" outer diameter. This fitting provides a quick and easy way to connect polyethylene tubing to threaded NPT ports. It can handle a variety of fluids including air, water, and chemicals. The push-to-connect design allows tubing to be securely connected without requiring clamps or threads.