Learn More
Grainger Connector 125VAC 15A L5-15R 2P 3W 1PH
Catalog No. 502764932
Encompass_Preferred
The Grainger Approved CL515DF Connector is a 125VAC 15A L5-15R 2P 3W 1PH connector. It has a NEMA configuration of L5-15R and is rated for 125 volts AC at 15 amps. The connector has 2 poles and 3 wires. It is designed for single phase power applications. The connector body is made of thermoplastic and has a nylon shell. It can be panel mounted and has screw flange terminals. The connector meets UL and CSA standards.
Catalog No. 50-276-4932 Supplier Grainger Supplier No. 16R093
The Grainger Approved CL515DF Connector is a 125VAC 15A L5-15R 2P 3W 1PH connector. It has a NEMA configuration of L5-15R and is rated for 125 volts AC at 15 amps. The connector has 2 poles and 3 wires. It is designed for single phase power applications. The connector body is made of thermoplastic and has a nylon shell. It can be panel mounted and has screw flange terminals. The connector meets UL and CSA standards.