Learn More
Grainger Connector 3/4 CTS x 3/4 NPT White
Catalog No. 502767286
Encompass_Preferred
The Grainger Approved 3/4" CTS x 3/4" NPT Brass Connector is made of brass and has a maximum pressure rating of 600 PSI at 70°F. It connects copper tubing to threaded pipe and can be used for air, water, oil, natural gas, and steam applications. The connector has a hex nut for tightening and is NSF/ANSI 61 certified for potable water service. It meets ASME B1.20.1 standards and is manufactured in the USA. The connector comes with a limited lifetime warranty.
Catalog No. 50-276-7286 Supplier Grainger Supplier No. 16T760
The Grainger Approved 3/4" CTS x 3/4" NPT Brass Connector is made of brass and has a maximum pressure rating of 600 PSI at 70°F. It connects copper tubing to threaded pipe and can be used for air, water, oil, natural gas, and steam applications. The connector has a hex nut for tightening and is NSF/ANSI 61 certified for potable water service. It meets ASME B1.20.1 standards and is manufactured in the USA. The connector comes with a limited lifetime warranty.