Learn More
MSC Mitutoyo Spc Cable, Left Connector, 1 m, 1 ea
Catalog No. 5000515705
Encompass
This SPC cable is designed for connecting various measuring instruments to data processors for efficient data transfer. It is compatible with a range of gages and units to facilitate accurate statistical process control in industrial environments.
  • Compatible with durometers, Hardmatic HH-300, absolute digimatic scale units, and digimatic miniprocessors
  • Suitable for use with calipers
  • Features a 1,016 mm (40 in.) cable length
  • Designed with a left-angle connector and 10-pin input plug for data processors
  • Compatible with all calipers without an absolute encoder
Catalog No. 50-005-15705 Supplier MSC Supplier No. 99580151
This SPC cable is designed for connecting various measuring instruments to data processors for efficient data transfer. It is compatible with a range of gages and units to facilitate accurate statistical process control in industrial environments.
  • Compatible with durometers, Hardmatic HH-300, absolute digimatic scale units, and digimatic miniprocessors
  • Suitable for use with calipers
  • Features a 1,016 mm (40 in.) cable length
  • Designed with a left-angle connector and 10-pin input plug for data processors
  • Compatible with all calipers without an absolute encoder