Learn More
AVANTOR MASTERFLEX (VWR LLC) CONNECTOR 5/8FTFT X 1/2FTFT 10
Catalog No. 502742394
Encompass
Masterflex Fitting, Natural Polypropylene, Straight, Hosebarb Reducer, 5/8IN ID x 1/2IN ID; 10/PK, Avantor
Catalog No. 50-274-2394 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061016
Masterflex Fitting, Natural Polypropylene, Straight, Hosebarb Reducer, 5/8IN ID x 1/2IN ID; 10/PK, Avantor