Learn More
Ace Glass, Inc. Connector, Ace-Safe, #11 to 3/8inid tubing, FETFE; O-ring
SDP
Supplier:  Ace Glass, Inc. 585810
Encompass
Ace-Safe threaded tubing connector connects #11 Ace-Threds to 3/8" I.D. tubing. A safety and flexibility improvement over glass hose connections. These connections dont break like glass, as they are manufactured from glass-filled PTFE for mechanical strength and chemical inertness. Stronger than PFA and polypropylene connectors. Supplied with a FETFE o-ring.
Catalog No. 50-972-383