Learn More
Ace Glass, Inc. Connector, Ace-Safe, #15 to 3/8inid tubing
SDP
Supplier:  Ace Glass, Inc. 585323
Encompass_Preferred
Ace-Safe polypropylene tubing connector, used to connect 3/8 I.D. flexible tubing to #15 Ace-Threds, for easy, safe connect/disconnect. Includes silicone O-ring and nylon bushing which are used to make a compression seal. Maximum temperature is 110 degrees C.
Catalog No. 50-978-525