Learn More
World Precision Instrument Connector, adapts WPI Transducers to non-WPI equipment
SDP
Catalog No. 50822064
Encompass
Need to use WPI transducers with non-WPI equipment?
This connector does the trick, with a female DIN connection on one end and an unwired connection on the other. This connector has no cable.
Catalog No. 50-822-064 Supplier World Precision Instrument Supplier No. 3492
Need to use WPI transducers with non-WPI equipment?
This connector does the trick, with a female DIN connection on one end and an unwired connection on the other. This connector has no cable.