Learn More
Chemglass Life Sciences Connector, Hook
SDP
Catalog No. 50871985
Encompass_Preferred
Connector, Hook. Easy-to-use connector allows one-handed assembly of two components with one adjustment screw. Maximum diameter of 1/2” (13mm) for each component.
Catalog No. 50-871-985 Supplier Chemglass Life Sciences Supplier No. CG926201
Connector, Hook. Easy-to-use connector allows one-handed assembly of two components with one adjustment screw. Maximum diameter of 1/2” (13mm) for each component.